Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15 Antibody (MM0368-13S34), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211665MFV610
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Western Blot, NeutralizationSpecifications
IL-15 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Neutralization | |
MM0368-13S34 | |
Western Blot, Block/Neutralize | |
IL-15MGC9721, interleukin 15, interleukin-15 | |
Human recombinant IL-15 | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
3600 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction