Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15 Antibody (OTI2B4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271034AF750
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Western Blot.Specifications
IL-15 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
IL15 | |
Recombinant protein fragment corresponding to amino acids 49-162 of human IL-15 (NP_751914) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2B4 | |
Western Blot | |
IL-15MGC9721, interleukin 15, interleukin-15 | |
Mouse | |
0.1 mL | |
3600 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction