Learn More
Invitrogen™ IL-15 Monoclonal Antibody (34559), PerCP

Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2, recombinant mouse IL-15, or rhIL-21 is observed.
Specifications
Specifications
| Antigen | IL-15 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 34559 |
| Conjugate | PerCP |
| Formulation | Buffered saline solution with 0.5% BSA and 0.09% sodium azide |
| Gene | IL15 |
| Gene Accession No. | P40933 |
| Gene Alias | AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15 |
| Gene Symbols | IL15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.