Learn More
IL-15 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 284551AP150UL
Description
The IL-15 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IL-15. This antibody recognizes human antigen. The IL-15 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
IL-15 | |
Polyclonal | |
Unconjugated | |
150 μL | |
3600 | |
IgG |
Immunohistochemistry, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.