Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Antibody (2639B), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10900C
This item is not returnable.
View return policy
Description
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Human samples. It is validated for Flow CytometrySpecifications
| IL-15R alpha | |
| Monoclonal | |
| PerCP | |
| CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Mouse myeloma cell line, NS0-derived human IL-15R alpha, Met1-Lys173, Accession # NP_751951 | |
| 0.1 mL | |
| Cancer | |
| 3601 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 2639B | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction