Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Antibody (888220), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5511PECY7
Description
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Mouse samples. It is validated for Flow CytometrySpecifications
IL-15R alpha | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
888220 | |
Flow Cytometry | |
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-15 R alpha , Gly33-Lys205, Accession # Q60819 | |
0.1 mL | |
Cancer | |
3601 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction