Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241212
Description
IL-15R alpha Polyclonal specifically detects IL-15R alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
IL-15R alpha | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
IL15RA | |
Antibody was raised against a 16 amino acid peptide near the center of human IL-15RA. The immunogen is located within amino acids 40 - 90 of IL-15RA. Amino Acid Squence: NSGFKRKAGTSSLTE | |
Affinity Purified | |
RUO | |
3601 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot, ELISA | |
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
Rabbit | |
30 kDa | |
0.1 mg | |
Primary | |
Multiple isoforms of IL-15RA are known to exist. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction