Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Antibody (JM7A4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143238AF350
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-15R alpha | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
IL15RA | |
The immunogen for this antibody was IL15Ra. | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry | |
JM7A4 | |
Western Blot, Flow Cytometry | |
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
Mouse | |
0.1 mL | |
3601 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction