Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-16 Antibody (70720), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3161L
Description
IL-16 Monoclonal antibody specifically detects IL-16 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
IL-16 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
70720 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
E. coli-derived recombinant human IL-16 isoform 1, Met1203-Ser1332, Accession # Q14005 | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity | |
3603 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction