Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-16 Chimeric Recombinant Rabbit Monoclonal Antibody (14.1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548123
Description
Specificity: The antibody is specific for IL-16.
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
Specifications
IL-16 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Q14005 | |
IL16 | |
The antibody was raised against IL-16 using a synthetic peptide corresponding to the predicted amino acid sequence of a hydrophilic region of IL-16, conjugated to keyhole limpet hemocyanin. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
14.1 | |
Unconjugated | |
IL16 | |
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 | |
Rabbit | |
Protein A | |
RUO | |
3603 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction