Learn More
Invitrogen™ IL-16 Monoclonal Antibody (17.1)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ M160E
Catalog No. ENM160E
Only null left
Edge
Add to Cart
Description
Description
M160E targets IL-16 in ELISA, and WB applications and shows reactivity with Human samples. The M160E immunogen is recombinant human IL-16. M160E detects IL-16 which has a predicted molecular weight of approximately 142 kDa. The M160E IL16 antibody (clone 17.1) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M161B (biotinylated conjugate of clone 14.1) Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Antibody M160E (clone 17.1) and biotinylated antibody M161B (clone 14.1) have successfully been used in combination with recombinant IL16 protein SIL16 in ELISA applications.
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-16
Monoclonal
1 mg/mL
PBS with no preservative
Q14005
IL16
Recombinant human IL-16.
500 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
17.1
Unconjugated
IL16
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
Mouse
Protein G
RUO
3603
-20°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.