Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody (408H6.01/mAb25), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0336B100
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Block/Neutralize.Specifications
IL-17/IL-17A | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry, ELISA 1:100-1:2000, Block/Neutralize | |
IL17A | |
IL17A was used as immunogen. | |
0.1 mg | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Store at -20C. Avoid freeze-thaw cycles. |
Blocking Assay, ELISA | |
408H6.01/mAb25 | |
Biotin | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction