Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody (41809) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB317J
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human, Primate samples. It is validated for Immunohistochemistry, Immunoprecipitation, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)Specifications
IL-17/IL-17A | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Immunohistochemistry, Immunoprecipitation, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
41809 | |
Immunohistochemistry, Immunoprecipitation, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
E. coli-derived recombinant human IL-17, Ile20-Ala155, Accession # Q16552 | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction