Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Antibody (4H1524), Novus Biologicals™
SDP

Catalog No. NBP227338SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227338SS 0.025 mg
NBP227338 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227338SS Supplier Novus Biologicals Supplier No. NBP227338SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot, Flow Cytometry, Block/Neutralize.

Specifications

Antigen IL-17/IL-17A
Applications Western Blot, Flow Cytometry, Blocking Assay
Classification Monoclonal
Clone 4H1524
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3ug/ml∼, Flow Cytometry 1:10-1:1000, Blocking/Neutralizing 0.025-0.25ug/ml
Gene Accession No. Q16552
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Gene Symbols IL17A
Host Species Mouse
Immunogen A portion of amino acids 1-100 was used as immunogen
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3605
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.