Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody (881309), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7211S
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-17/IL-17A | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
881309 | |
Flow Cytometry, CyTOF-ready | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
E. coli-derived recombinant mouse IL-17/IL-17A, Thr22-Ala158, Accession # Q62386 | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction