Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP142772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142772 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142772 Supplier Novus Biologicals Supplier No. NBP142772
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

IL-17/IL-17A Polyclonal specifically detects IL-17/IL-17A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-17/IL-17A
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000-1:5000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Gene Accession No. Q61453
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Gene Symbols IL17A
Host Species Rabbit
Immunogen This purified IL-17/IL-17A Antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant rat IL-17/IL-17A protein. (Uniprot: Q61453)
Molecular Weight of Antigen 16.88 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3605
Test Specificity In ELISA and other immunoreactive assays, this antibody will recognize both native and recombinant rat IL17-A in cell supernatants and certain body fluids. A control of similarly diluted normal rabbit IgG is recommended.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Human, Mouse, Rat
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.