Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP176337
Description
IL-17/IL-17A Polyclonal specifically detects IL-17/IL-17A in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
IL-17/IL-17A | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rhesus Macaque | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.125-2 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 5-20 ug/ml, Immunohistochemistry-Paraffin 1-5 ug/ml | |
NP_002181 | |
IL17A | |
Antibody was raised against a peptide corresponding to 16 amino acids near the center of mature human IL-17. The immunogen is located within amino acids 50-100 of IL-17. | |
0.1 mg | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction