Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody (MM17F3), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312035R
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
IL-17/IL-17A | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG κ |
Flow Cytometry, ELISA | |
MM17F3 | |
Flow Cytometry, ELISA | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
This antibody was raised by immunising C57Bl/6 mice with IL-17/IL-17A cross-linked to ovalbumin to generate a highly immunogenic complex. | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction