Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody (TC11-18H10), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP172027JF646
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-17/IL-17A | |
Monoclonal | |
0.5 mg/ml | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Rat | |
Affinity Purified | |
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Mouse | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
TC11-18H10 | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL17A | |
E. coli-expressed, recombinant mouse IL-17/IL-17A | |
0.1 mL | |
Primary | |
Recognizes mouse Interleukin 17, a 32kDa disulfide-linked homodimer consisting of two 136 amino acid chains, which belong to the IL-17 family (classified as IL-17A to IL-17F), and signal through the IL-17 receptor (IL-17R/CDw217). | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction