Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Antibody (TC11-18H10), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP172027PEATT594
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-17/IL-17A | |
Monoclonal | |
0.5 mg/ml | |
Flow Cytometry | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Rat | |
Affinity Purified | |
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
3605.0 | |
Mouse | |
IgG1 |
Flow Cytometry | |
TC11-18H10 | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
IL17A | |
E. coli-expressed, recombinant mouse IL-17/IL-17A | |
0.1 mL | |
Primary | |
Recognizes mouse Interleukin 17, a 32kDa disulfide-linked homodimer consisting of two 136 amino acid chains, which belong to the IL-17 family (classified as IL-17A to IL-17F), and signal through the IL-17 receptor (IL-17R/CDw217). | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction