Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Mouse anti-Human, Alexa Fluor™ 532, Clone: 4k5F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP224765AF532

Catalog No. NP22476A32


Add to cart

Description

Description

IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-17/IL-17A
4k5F6
Western Blot
Conjugated, Purified
IL17A
A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Protein G purified
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary
3605.0
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit