Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17 Receptor B Rat anti-Mouse, Brilliant Violet 421, Clone: 6B7, BD Optibuild™

Catalog No. BDB743815 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743815 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743815 Supplier BD Biosciences Supplier No. 743815
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6B7 monoclonal antibody specifically binds to IL-17 Receptor B (IL-17RB), which is a single-pass type I transmembrane protein and a component of the heterodimeric receptor for IL-25 (also known as IL-17E). Several components of the IL-17RB signaling mechanism have been identified, including ACT1, TRAF4, SMURF2, DAZAP2, and STAT5. The cytoplasmic tails of both of the IL-25 Receptor components, IL-17RA and IL-17RB, and ACT1 contain the SEFIR (similar expression to fibroblast growth factor genes and IL-17R) domain that is essential for IL-25R-mediated signaling. Signaling through IL-17RB mediates Th2 immune responses and is responsible for pathogenesis in a murine model for asthma. IL-17RB is found on subsets of iNKT cells, Th2 cells, some myeloid cells in solid organs, and type 2 innate lymphoid cells (ILC2) in lung, peritoneum, mesenteric lymph nodes and bone marrow.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17 Receptor B
Classification Monoclonal
Clone 6B7
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IL-17RB, IL-17B receptor, Il17rb, Evi27, Il17rb, IL-17ER, IL17RH1, IL-17Rh1
Host Species Rat
Immunogen Mouse IL-17 Receptor B Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.