Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17A Monoclonal Antibody (eBio64CAP17), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$2500.00 - $12337.65
Specifications
Antigen | IL-17A |
---|---|
Clone | eBio64CAP17 |
Concentration | 1 mg/mL |
Classification | Monoclonal |
Conjugate | Functional Grade |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-717-8050
![]() |
Invitrogen™
167178050 |
50 mg |
Each
for $12,337.65
Save 25% Reg : |
|
|||||
50-255-0874
![]() |
Invitrogen™
16717838 |
5 mg |
Each for $2,500.00
|
|
|||||
Description
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Specifications
IL-17A | |
1 mg/mL | |
Functional Grade | |
Mouse | |
Human | |
3605 | |
Primary | |
Antibody |
eBio64CAP17 | |
Monoclonal | |
Liquid | |
RUO | |
Q16552 | |
IgG1 κ | |
Affinity Chromatography | |
IL17A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title