Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17A, Mouse Anti-Human, RB613, Clone: N49-653, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 571130
Description
IL-17A Monoclonal antibody specifically detects IL-17A in Human (QC Testing) samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
IL-17A | |
Monoclonal | |
RB613 | |
IL17; IL-17; CTLA8; Cytotoxic T-lymphocyte-associated serine esterase 8 | |
Human IL-17A Recombinant Protein | |
100 Tests | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining) | |
N49-653 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
RUO | |
3605 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction