Learn More
IL-17A Polyclonal Antibody, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
$329.00
Specifications
| Antigen | IL-17A |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P07G100
|
PeproTech
500P07G100UG |
100 μg |
Each of 1 for $329.00
|
|
|||||
Description
AA Sequence of recombinant protein: MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-17A. Anti-Human IL-17A-specific antibody was purified by affinity chromatography employing an immobilized Human IL-17 matrix. Sandwich ELISA: To detect Human IL-17A by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-17A (500-P07Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-17A. Western Blot: To detect Human IL-17A by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-17A is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Specifications
| IL-17A | |
| ELISA, Western Blot | |
| Unconjugated | |
| Goat | |
| Human | |
| Q16552 | |
| 3605 | |
| E.coli-derived Recombinant Human IL-17A. | |
| Antigen affinity chromatography | |
| IL17A |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A | |
| IL17A | |
| Primary | |
| -20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.