Learn More
Invitrogen™ IL-17AF Monoclonal Antibody (20LJS09), PE, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 12917942

Description
Description: The 20LJS09 monoclonal antibody reacts with human interleukin (IL)-17AF heterodimer. This antibody specifically reacts with this heterodimer and has no reactivity for IL-17A or IL-17F homodimers. IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis. Applications Reported: This 20LJS09 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 20LJS09 antibody has been pre-titrated and tested on 10 day in vitro-polarized Th17 cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.
Specifications
IL-17AF | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q16552, Q96PD4 | |
Il17a, Il17f | |
Affinity chromatography | |
RUO | |
112744, 3605 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
20LJS09 | |
PE | |
Il17a | |
C87042; CANDF6; Ctla8; CTLA-8; cytokine ML-1; Cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; IL17; IL-17; Il17a; IL-17A; IL17F; IL-17F; ILN; Interleukin; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17A; interleukin 17F; interleukin-17A; Interleukin-17F; ML1; ML-1 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.