Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17B Antibody (B-B58), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318115AF405
Description
IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for ELISASpecifications
IL-17B | |
Monoclonal | |
Alexa Fluor 405 | |
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
Recombinant human IL-17B. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
27190 | |
Store at 4°C in the dark. | |
IgG2a κ |
ELISA | |
B-B58 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction