Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17C Antibody (311522), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB23061Y
Description
IL-17C Monoclonal antibody specifically detects IL-17C in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
IL-17C | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
311522 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
E. coli-derived recombinant mouse IL-17C, His15-Gln194, Accession # NP_665833 | |
0.1 mL | |
Cytokine Research | |
27189 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction