Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17C Antibody (311522), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB23061MFV610
Description
IL-17C Monoclonal antibody specifically detects IL-17C in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| IL-17C | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 311522 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
| E. coli-derived recombinant mouse IL-17C, His15-Gln194, Accession # NP_665833 | |
| 0.1 mL | |
| Cytokine Research | |
| 27189 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction