Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17D Mouse anti-Human, Clone: 1D7H4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | IL-17D |
---|---|
Clone | 1D7H4 |
Concentration | 1.33 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6578
![]() ![]() |
Proteintech Group Inc
661431IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.Specifications
IL-17D | |
1.33 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IL 17D, IL 27, IL17D, IL27, interleukin 17D, Interleukin 27 | |
IL17D | |
IgG2b | |
Protein A | |
Antibody |
1D7H4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8TAD2 | |
53342 | |
IL-17D Fusion Protein Ag6762 | |
Primary | |
-20°C | |
IL17D |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title