Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17E/IL-25 Antibody (110), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290448V
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for ELISA.Specifications
IL-17E/IL-25 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
110 | |
ELISA | |
IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-17E/IL-25 (NP_542767.1; Val 17-Ala 169). | |
0.1 mL | |
Cytokine Research | |
64806 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction