Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17E/IL-25 Antibody (182203), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1258G
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, ELISA, Flow Cytometry, CyTOFSpecifications
IL-17E/IL-25 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Flow Cytometry, CyTOF | |
182203 | |
Western Blot, ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
E. coli-derived recombinant human IL-17E, Tyr33-Gly177, Accession # Q9H293 | |
0.1 mL | |
Cytokine Research | |
64806 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction