Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17E/IL-25 Antibody (207702) [PerCP], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB13991C
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Mouse samples. It is validated for ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOFSpecifications
IL-17E/IL-25 | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF | |
207702 | |
ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready | |
IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
E. coli-derived recombinant mouse IL-17E/IL-25, Val17-Ala169, Accession # NP_542767 | |
0.1 mL | |
Cytokine Research | |
64806 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction