Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17E/IL-25 Antibody (68C1039.2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056541B
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-17E/IL-25 | |
Monoclonal | |
Biotin | |
Q9H293 | |
IL25 | |
Amino acids 115-132 of human IL-17E (isoform CRA_a) were used as immunogen for this antibody. | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
68C1039.2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
Mouse | |
20 kDa | |
0.1 mL | |
Cytokine Research | |
64806 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction