Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17E/IL-25 Antibody (8B1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227324R
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IL-17E/IL-25 | |
Monoclonal | |
DyLight 550 | |
Q9H293 | |
IL25 | |
Full-length human IL17E recombinant protein was used as the immunogen for this antibody. | |
0.1 mL | |
Cytokine Research | |
64806 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry | |
8B1 | |
Western Blot, Flow Cytometry | |
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction