Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17F Antibody (003), Alexa Fluor™ 532

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290005AF532
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for ELISA.Specifications
IL-17F | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
3 | |
ELISA | |
Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-17F (NP_443104.1; Arg31-Gln163). | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
112744 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction