Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    IL-17F Antibody (1058A), HRP, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB20571H
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow CytometrySpecifications
| IL-17F | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4C in the dark. | 
| Flow Cytometry | |
| 1058A | |
| Flow Cytometry | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| E.coli-derived recombinant mouse IL-17F, Ala21-Val153, Accession # Q7TNI7 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction