Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17F Antibody (197301), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB13351T
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
IL-17F | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
197301 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
E. coli-derived recombinant human IL-17F, Arg31-Gln163, Accession # Q96PD4 | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
112744 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction