Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17F Antibody (316016), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2057K
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
IL-17F | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
316016 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
E. coli-derived recombinant mouse IL-17F, Arg29-Ala161, Accession # Q7TNI7 | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
112744 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction