Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17F Antibody (4H450) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7110691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227319SS 0.025 mg
NBP227319 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227319SS Supplier Novus Biologicals Supplier No. NBP227319SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-17F
Applications Western Blot
Classification Monoclonal
Clone 4H450
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.2 - 2 ug/ml
Formulation Sterile - filtered PBS with No Preservative
Gene Accession No. Q96PD4
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24
Gene Symbols IL17F
Host Species Mouse
Immunogen Full-length recombinant MRkIPkVGHT FFQkPESCPP VPGGSMkLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGkEDISMN SVPIQQETLV VRRkHQGCSV SFQLEkVLVT VGCTCVTPVI HHVQ
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.