Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17F Antibody (8F5.1A9), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312034JF646
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow CytometrySpecifications
IL-17F | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
8F5.1A9 | |
Flow Cytometry | |
Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
This antibody was raised by immunising mice with murine IL-17F coupled to ovalbumin to generate a highly immunogenic complex. | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
112744 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction