Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17F Antibody (B-F60), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314589C
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for ELISASpecifications
IL-17F | |
Monoclonal | |
DyLight 650 | |
Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
Recombinant human IL-17F | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
112744 | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA | |
B-F60 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction