Learn More
IL-17f Monoclonal Antibody (316016)

Description
IL-17f Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | IL-17f |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 316016 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | IL17F |
| Gene Accession No. | Q7TNI7 |
| Gene Alias | C87042; CANDF6; cytokine ML-1; IL17F; IL-17F; IL-24; ILN; Interleukin; interleukin 17F; Interleukin17F; interleukin-17F; interleukin-24; ML1; ML-1; mutant IL-17F |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.