Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17F Rabbit anti-Mouse, PE, Clone: 1058A, R&D Systems™

Catalog No. IC20571P025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC20571P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC20571P025 Supplier R&D Systems Supplier No. IC20571P025
This item is not returnable. View return policy

Recombinant Monoclonal Antibody

IL-17F, also called ML-1, is a 21-33kDa member of the IL-17 family of protein. It is one of six glycoproteins with conserved cysteines that form a cystine knot structure. IL-17F is expressed by multiple cell types, including bronchial epthelium, neutrophils (likely), monocytes, mast cells, colonic epithelium, γδ T cells, CD4+ Th17 cells, group 3 (NKp46- RORγt) innate lymphoid cells, NKT cells and CD8+ Tc17 cells. IL-17F is known to form disulfide-linked homodimers, and covalent heterodimes with IL-17A. Notable, the IL-17A and IL-17F homodimers, plus the IL-17A:F heterodimer use the same receptor combination (IL-17RA:IL-17RC), but elicit somewhat different outcomes. For instance, on macrophages, IL-17A induces IL-9, KC and GM-CSF secretion, while IL-17F does not. Both molecules, however, induce the secretion of the same molecules (IL-9, KC, and GM-CSF) by colonic epithelium. Mouse IL-17F has a long form and a short form, which have alternate start sites but identical lengths. Over amino acids (aa) 21-153, mouse IL-17F shares 88% and 56% aa sequence identity with rat and human IL-17F, respectively.

Specifications

Antigen IL-17F
Applications Flow Cytometry
Classification Monoclonal
Clone 1058A
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q7TNI7
Gene Alias Cytokine ML-1, IL17F, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24
Gene Symbols Il17f
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse IL-17F Ala21-Val153 Accession # Q7TNI7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112744
Test Specificity Detects mouse IL-17F in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.