Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RA/IL-17R Antibody (012), DyLight 550 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289700R
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for ELISASpecifications
IL-17RA/IL-17R | |
Monoclonal | |
DyLight 550 | |
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-17RA/IL-17R (NP_055154.3; Met 1-Trp 320). | |
0.1 mL | |
Innate Immunity | |
23765 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
012 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction