Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RA/IL-17R Antibody (49M4D2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
Antigen | IL-17RA/IL-17R |
---|---|
Clone | 49M4D2 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225258SS
![]() |
Novus Biologicals
NBP225258SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP225258
![]() |
Novus Biologicals
NBP225258 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-17RA/IL-17R | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
IL17RA | |
IgG1 κ | |
Protein G purified | |
This product reacts with Human. |
49M4D2 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
NP_055154 | |
23765 | |
Within the range of amino acids 186-267 of human IL-17RA were used as the immunogen for the antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title