Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RA/IL-17R Antibody (657603), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4481X
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
IL-17RA/IL-17R | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Neutralization, CyTOF | |
657603 | |
Flow Cytometry, Neutralization, CyTOF-ready | |
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RA/IL-17 R, Ser32-Trp322, Accession # Q60943 | |
0.1 mL | |
Innate Immunity | |
23765 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction