Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RA/IL-17R Antibody (CL10283), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | IL-17RA/IL-17R |
---|---|
Clone | CL10283 |
Dilution | Western Blot 1 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western BlotSpecifications
IL-17RA/IL-17R | |
Western Blot 1 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
This antibody was developed against a Synthetic Peptide of Q96F46. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL10283 | |
Western Blot | |
Unconjugated | |
Mouse | |
Innate Immunity | |
PBS, pH 7.2, 40% glycerol | |
23765 | |
IgG2b | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title