Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RA/IL-17R Antibody (Hu129), PE/Cy7, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB10547PECY7
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Flow CytometrySpecifications
IL-17RA/IL-17R | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
Hu129 | |
Flow Cytometry | |
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
Human Il-17 Receptor | |
0.1 mL | |
Innate Immunity | |
23765 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction