Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RB Antibody (938923) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | IL-17RB |
---|---|
Clone | 938923 |
Concentration | 1.0 mg/mL |
Dilution | Immunohistochemistry 10 - 15 ug/ml, Immunohistochemistry-Paraffin 10 - 15 ug/ml, Immunocytochemistry 5 - 10 ug/ml |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12073SP
![]() |
Novus Biologicals
MAB12073SP |
25 μg |
Each for $181.90
|
|
|||||
MAB12073
![]() |
Novus Biologicals
MAB12073 |
100 μg |
Each for $487.50
|
|
|||||
Description
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IL-17RB | |
1.0 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Mouse | |
Cytokine Research | |
PBS with No Preservative | |
55540 | |
Partial recombinant human IL-17RB protein (aa18-289) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
938923 | |
Immunohistochemistry 10 - 15 ug/ml, Immunohistochemistry-Paraffin 10 - 15 ug/ml, Immunocytochemistry 5 - 10 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, EVI27IL-17B receptor, IL-17 receptor B, IL17BRinterleukin 17B receptor, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
IL17RB | |
IgG2b | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title